Learn More
Bellco Glass, Inc. GL45 6 Baffle Shake Flask,2000mL With Membrane Screw Cap
SDP
Catalog No. 501945545
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. 3 Side baffles positioned at 120 degree and 3 bottom baffles offset from the side baffles. GL45 polypropylene membrane cap has a sealed in 0.2u PTFE membrane. Cap can be tightened during autoclaving.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassIncludes Polypropylene Membrane Cap And Pouring RingSize: 2LDimensions: 158x260mmCase Quantity: 3
Catalog No. 50-194-5545 Supplier Bellco Glass, Inc. Supplier No. 254502000
Manufactured from Type 1 Class A Borosilicate Glass. 3 Side baffles positioned at 120 degree and 3 bottom baffles offset from the side baffles. GL45 polypropylene membrane cap has a sealed in 0.2u PTFE membrane. Cap can be tightened during autoclaving.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassIncludes Polypropylene Membrane Cap And Pouring RingSize: 2LDimensions: 158x260mmCase Quantity: 3