Learn More
Ace Glass, Inc. GL45 Cap with 10/30 Outer Joint Yellow PP PTFE 10/30 joint fixed in GL45 Yellow Polypropylene cap with machined groove for Keck clips
SDP
Catalog No. 5000810365
Encompass
GL45 Cap with 10/30 Outer Joint Yellow PP PTFE 10/30 joint fixed in GL45 Yellow Polypropylene cap with machined groove for Keck clips
Catalog No. 50-008-10365 Supplier Ace Glass, Inc. Supplier No. 522413
GL45 Cap with 10/30 Outer Joint Yellow PP PTFE 10/30 joint fixed in GL45 Yellow Polypropylene cap with machined groove for Keck clips