Learn More
Ace Glass, Inc. GL45 Cap with 14/20 Outer Joint Orange PP PTFE 14/20 joint fixed in GL45 Orange Polypropylene cap with machined groove for Keck clips
SDP
Catalog No. 5000810378
Encompass
GL45 Cap with 14/20 Outer Joint Orange PP PTFE 14/20 joint fixed in GL45 Orange Polypropylene cap with machined groove for Keck clips
Catalog No. 50-008-10378 Supplier Ace Glass, Inc. Supplier No. 522435
GL45 Cap with 14/20 Outer Joint Orange PP PTFE 14/20 joint fixed in GL45 Orange Polypropylene cap with machined groove for Keck clips