Learn More
Ace Glass, Inc. GL45 Cap with 29/32 Outer Joint Blue PP PTFE 29/32 joint fixed in GL45 Blue Polypropylene cap with machined groove for Keck Clips
SDP
Catalog No. 5000810377
Encompass
GL45 Cap with 29/32 Outer Joint Blue PP PTFE 29/32 joint fixed in GL45 Blue Polypropylene cap with machined groove for Keck Clips
Catalog No. 50-008-10377 Supplier Ace Glass, Inc. Supplier No. 522412
GL45 Cap with 29/32 Outer Joint Blue PP PTFE 29/32 joint fixed in GL45 Blue Polypropylene cap with machined groove for Keck Clips