Learn More
Bellco Glass, Inc. GL45 L-Bubble Tube W/Holes,36L With (10) .040 Holes"
SDP
Catalog No. 501944795
Encompass_Preferred
Sparger for vertical sidearm spinner flasks. Includes polypropylene cap with 1/4" compression fitting with .040" holes for fermentation where shear is not critical and white silicone gasket. MADE IN THE USA. Material: 316l stainless steel with electropolish finish Ra 32 finish or better Includes polypropylene cap, 1/4" white compression fitting and white silicone gasket. DIMENSIONS: 21 X 7 1/4"
Catalog No. 50-194-4795 Supplier Bellco Glass, Inc. Supplier No. 196436145
Sparger for vertical sidearm spinner flasks. Includes polypropylene cap with 1/4" compression fitting with .040" holes for fermentation where shear is not critical and white silicone gasket. MADE IN THE USA. Material: 316l stainless steel with electropolish finish Ra 32 finish or better Includes polypropylene cap, 1/4" white compression fitting and white silicone gasket. DIMENSIONS: 21 X 7 1/4"