Learn More
Bellco Glass, Inc. GL45 L-Bubbler Tube Assembly ,36L W/Stainless Steel Frit - For Over Head Drive Vessels
SDP
Catalog No. 501944794
Encompass_Preferred
Sparger for vertical sidearm spinner flasks. Includes polypropylene cap with 1/4" compression fitting and white silicone gasket. MADE IN THE USA. Material: 316l stainless steel with electropolish finish Ra 32 finish or better Size: 21"
Catalog No. 50-194-4794 Supplier Bellco Glass, Inc. Supplier No. 196436045
Sparger for vertical sidearm spinner flasks. Includes polypropylene cap with 1/4" compression fitting and white silicone gasket. MADE IN THE USA. Material: 316l stainless steel with electropolish finish Ra 32 finish or better Size: 21"