Learn More
Description
- 33 Expansion Borosilicate Glass
- Drip Free
- Colored Cap
- No Seam
Specifications
Specifications
| Capacity (Metric) | 250 mL |
| Color | Clear |
| Material | Borosilicate Glass (Class A, Type I) |
| Quantity | 1 Ea. |
| Sterility | Non-sterile |
| Closure Color | Yellow |
| Closure Included | Yes |
| Graduated | Yes |
| Mouth | Wide, Threaded |
| Fit for Purpose | Certified Quality |
| Show More |
Caps must be removed for autoclaving!
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

