Learn More
Aviva Systems Biology Corp GLA Antibody - N-terminal region (ARP54296_P050) | 100uL
SDP
Catalog No. 5001461165
Encompass_Preferred
This is a rabbit polyclonal antibody against GLA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GLA. Alternate names: GALAApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61165 Supplier Aviva Systems Biology Corp Supplier No. ARP54296_P050
This is a rabbit polyclonal antibody against GLA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GLA. Alternate names: GALAApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish