Learn More
Medchemexpress LLC Glabrol | 59870-65-4 | 5 MG
SDP
Catalog No. 5000420852
Encompass_Preferred
Glabrol is an isoprenyl flavonoid isolated from ethanol extract of licorice roots. It acts as a potent and non-competitive Acyl-coenzyme A: cholesterol acyltransferase (ACAT) inhibitor with an IC50 value of 24.6 μM for rat liver microsomal ACAT activity. This product is intended for research use only.
  • Isoprenyl flavonoid isolated from licorice roots
  • Potent and non-competitive acyl-coenzyme A: cholesterol acyltransferase (ACAT) inhibitor
  • Decreases cholesteryl ester formation in HepG2 cells
  • For research use only
Catalog No. 50-004-20852 Supplier Medchemexpress LLC Supplier No. HYN41935MG
Glabrol is an isoprenyl flavonoid isolated from ethanol extract of licorice roots. It acts as a potent and non-competitive Acyl-coenzyme A: cholesterol acyltransferase (ACAT) inhibitor with an IC50 value of 24.6 μM for rat liver microsomal ACAT activity. This product is intended for research use only.
  • Isoprenyl flavonoid isolated from licorice roots
  • Potent and non-competitive acyl-coenzyme A: cholesterol acyltransferase (ACAT) inhibitor
  • Decreases cholesteryl ester formation in HepG2 cells
  • For research use only