Learn More
Medchemexpress LLC Glabrol | 59870-65-4 | 99.8% | 392.49 g·mol⁻1 | C25H28O4 | 10 MG
SDP
Catalog No. 5000289523
Encompass_Preferred
Glabrol is an isoprenyl flavonoid research reagent that functions as a non-competitive inhibitor of acyl-coenzyme A: cholesterol acyltransferase (ACAT). It is intended for laboratory research use in biochemical assays and inhibitor characterization.
  • Isoprenyl flavonoid isolated from licorice root.
  • Non-competitive ACAT inhibitor with reported IC50 = 24.6 μM (rat liver microsomes).
  • C25H28O4; molecular weight 392.49 g·mol⁻1.
  • High purity (≈99.8%).
  • Supplied as a 10 mg research sample; store at 4°C and protect from light.
Catalog No. 50-002-89523 Supplier Medchemexpress LLC Supplier No. HYN419310MG
Glabrol is an isoprenyl flavonoid research reagent that functions as a non-competitive inhibitor of acyl-coenzyme A: cholesterol acyltransferase (ACAT). It is intended for laboratory research use in biochemical assays and inhibitor characterization.
  • Isoprenyl flavonoid isolated from licorice root.
  • Non-competitive ACAT inhibitor with reported IC50 = 24.6 μM (rat liver microsomes).
  • C25H28O4; molecular weight 392.49 g·mol⁻1.
  • High purity (≈99.8%).
  • Supplied as a 10 mg research sample; store at 4°C and protect from light.