Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glacial Acetic Acid Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111019166 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
3 x 1.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9166 3 x 1.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9166 Supplier MilliporeSigma Supelco Supplier No. PHR17483X1.5ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glacial Acetic Acid is a simple monocarboxylic acid, which is devoid of water. Pharmaceutically, it is commonly used as a topical antibiotic that treats infections caused by bacteria.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Featured Industries: Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 64-19-7
Ignition Point 800°F
Vapor Density 2.07 (vs. air)
Density 1.049 g/mL (at 25°C)
Flash Point 39°C
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula CH3 CO2 H
MDL Number MFCD00036152
Melting Point 16.2°C
Boiling Point 117°C to 118°C
Synonym Acetic acid; Glacial acetic acid
Type Certified Reference Material
Quantity 3 x 1.5 mL
Formula Weight 60.05
Grade Certified Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.