Learn More
Genesee Scientific Corporation GLAD PRESS'N SEAL PLAS ROLL
SDP
Catalog No. NC9287033
Encompass
Glad Press'n Seal, Glad press'n seal sealable plastic wrap to (plug) vials in the fly kitchen, Leaving the real plugging to be done when flies are on the vials, 140sq. ft., 1/RL
Catalog No. NC9287033 Supplier Genesee Scientific Corporation Supplier No. 88142
Glad Press'n Seal, Glad press'n seal sealable plastic wrap to (plug) vials in the fly kitchen, Leaving the real plugging to be done when flies are on the vials, 140sq. ft., 1/RL