Learn More
Perkin Elmer US LLC GladiATR Heated Diamond Crystal Plate - 210° C
Catalog No. 502113540
Encompass_Preferred
The GladiATR Vision optical design is all reflective, preserving the full spectral range inherent to diamond. For standard mid-IR, the spectral range available with the GladiATR Vision will be 400-400 cm-1. For Far-IR configuration, the spectral range is extended to less than 50 cm-1. The GladiATR Diamond ATR accessory is compatible with the Spectrum 3/Frontier spectrometers. It includes: Base Optics Monolithic diamond crystal plate High pressure clamp with three interchangeable tips Liquids retainer with volatiles cover Purge tubes Purge kit Spectrometer base mount
Catalog No. 50-211-3540 Supplier Perkin Elmer US LLC Supplier No. L1272422
Add to Cart
Add to Cart
Request a Quote
The GladiATR Vision optical design is all reflective, preserving the full spectral range inherent to diamond. For standard mid-IR, the spectral range available with the GladiATR Vision will be 400-400 cm-1. For Far-IR configuration, the spectral range is extended to less than 50 cm-1. The GladiATR Diamond ATR accessory is compatible with the Spectrum 3/Frontier spectrometers. It includes: Base Optics Monolithic diamond crystal plate High pressure clamp with three interchangeable tips Liquids retainer with volatiles cover Purge tubes Purge kit Spectrometer base mount