Learn More
Medchemexpress LLC Glafenine | 3820-67-5 | MFCD00079326 | 98.8% | 250 MG
SDP
Catalog No. 5000414406
Encompass_Preferred
Glafenine is a non-selective, non-steroidal anti-inflammatory drug-based COX-1/COX-2 inhibitor. It exhibits anti-inflammatory, anti-proliferative, and anti-cell migration effects by inhibiting the arachidonic acid metabolic pathway and reducing prostaglandin synthesis. It is useful in research for inflammatory-related diseases, vascular restenosis, and cystic fibrosis.
  • Induces cell cycle arrest in vascular smooth muscle cells and endothelial cells
  • Reduces the synthesis of the extracellular matrix protein tenascin
  • Can be utilized in research for inflammatory-related diseases, vascular restenosis, and cystic fibrosis (CF)
Catalog No. 50-004-14406 Supplier Medchemexpress LLC Supplier No. 250MGHYB1153
Glafenine is a non-selective, non-steroidal anti-inflammatory drug-based COX-1/COX-2 inhibitor. It exhibits anti-inflammatory, anti-proliferative, and anti-cell migration effects by inhibiting the arachidonic acid metabolic pathway and reducing prostaglandin synthesis. It is useful in research for inflammatory-related diseases, vascular restenosis, and cystic fibrosis.
  • Induces cell cycle arrest in vascular smooth muscle cells and endothelial cells
  • Reduces the synthesis of the extracellular matrix protein tenascin
  • Can be utilized in research for inflammatory-related diseases, vascular restenosis, and cystic fibrosis (CF)