Learn More
Crystal Chem Inc Glargine ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 90125
Encompass
The Glargine ELISA is a highly sensitive assay used to quantify levels of human glargine in plasma.Highlights: Uses 25 µL sample. Wide dyanamic range of 10 - 250 mU/L. High sensitivity of 0.8 mU/L.Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7928