Learn More
Glas Col LLC Heating Mantle
SDP
Catalog No. NC1445363
Encompass_Preferred
Fabric heating mantle for 22L speherical flask to 400 degrees C maximum internal operating temperature. 2@700W, 115V CSA
Catalog No. NC1445363 Supplier Glas Col LLC Supplier No. 100A O418
Fabric heating mantle for 22L speherical flask to 400 degrees C maximum internal operating temperature. 2@700W, 115V CSA