Learn More
Glas Col LLC Vortexer
SDP
Catalog No. 501616334
Encompass_Preferred
Large Capacity Mixer. The unit is designed to handle a wide variety of vessels from micro-centrifuge tubes to large vessels, such as Erlenmeyer flasks. The interchangeable cartridge system allows the user to quickly switch from one type of vessel to another.the pulsing feature interrupts vortexing to join upper and lower strata for more thorough mixing: range from 60 to 80 PPM. 100-2,000rpm, 1.5A.50/60Hz, 120V, max. load weight 5 lbs.
Catalog No. 50-161-6334 Supplier Glas Col LLC Supplier No. 099A LC1012
Large Capacity Mixer. The unit is designed to handle a wide variety of vessels from micro-centrifuge tubes to large vessels, such as Erlenmeyer flasks. The interchangeable cartridge system allows the user to quickly switch from one type of vessel to another.the pulsing feature interrupts vortexing to join upper and lower strata for more thorough mixing: range from 60 to 80 PPM. 100-2,000rpm, 1.5A.50/60Hz, 120V, max. load weight 5 lbs.