Learn More
Andwin Scientific Heating mantles
SDP
Catalog No. 5089990321
Encompass_Preferred
Heating mantles are preferred to Bunsen burners or hot plates for organic liquids. They provide even heating (no hot spots) and maximum surface coverage for better temperature uniformity. The Series O spherical flask mantle covers only the flask's bottom half, letting you see its full contents. It should be used with the Series O supports. Fabric heating mantle, 200ml, 100w
Catalog No. 50-899-90321 Supplier Andwin Scientific Supplier No. 100AO400
Heating mantles are preferred to Bunsen burners or hot plates for organic liquids. They provide even heating (no hot spots) and maximum surface coverage for better temperature uniformity. The Series O spherical flask mantle covers only the flask's bottom half, letting you see its full contents. It should be used with the Series O supports. Fabric heating mantle, 200ml, 100w