Learn More
Ace Glass, Inc. Glas-Col Rotator riser option lifts unit so larger glassware will clear the bench
SDP
Catalog No. 50978127
Encompass
Glas-Col Rotator riser option lifts unit so larger glassware will clear the bench
Catalog No. 50-978-127 Supplier Ace Glass, Inc. Supplier No. 1384640
Glas-Col Rotator riser option lifts unit so larger glassware will clear the bench