Learn More
Glas Col LLC Seperatory Funnel Glass
SDP
Catalog No. NC0622771
Encompass_Preferred
2L separatory funnel with PTFE cap and spout. For use with bench top, 3D midrange and 3D floor shakers.
Catalog No. NC0622771 Supplier Glas Col LLC Supplier No. 099A SEP2000
2L separatory funnel with PTFE cap and spout. For use with bench top, 3D midrange and 3D floor shakers.