Learn More
Glas Col LLC Heating Mantle
SDP
Supplier:  Glas Col LLC 100B TM102
Encompass_Preferred
Aluminum housed heating mantle 250ml round bottom flask to 400 degrees C maximum internal operating temperature.115V, 180W CSA. The Series TM mantle covers only the flask's bottom half, letting you see its full contents. Larger sizes have multiple circuits for ease of temperature control when a flask is less than half full. . All 115 volt units are CSA certified.Series TM mantles are available with a bottom hole for flask drains.
Catalog No. 50-161-6368