Learn More
Glas Col LLC Heating Mantle
SDP
Catalog No. 501616371
Encompass_Preferred
Aluminum housed heating mantle 1L round bottom flask to 400 degrees C maximum internal operating temperature.115V, 380W CSA. The Series TM mantle covers only the flask's bottom half, letting you see its full contents. Larger sizes have multiple circuits for ease of temperature control when a flask is less than half full. . All 115 volt units are CSA certified.Series TM mantles are available with a bottom hole for flask drains.
Catalog No. 50-161-6371 Supplier Glas Col LLC Supplier No. 100B TM108
 Restricted Item
Add to Cart
Add to Cart
Aluminum housed heating mantle 1L round bottom flask to 400 degrees C maximum internal operating temperature.115V, 380W CSA. The Series TM mantle covers only the flask's bottom half, letting you see its full contents. Larger sizes have multiple circuits for ease of temperature control when a flask is less than half full. . All 115 volt units are CSA certified.Series TM mantles are available with a bottom hole for flask drains.