Learn More
Glas Col LLC Heating Mantle
SDP
Catalog No. 501616364
Encompass_Preferred
Aluminum housed heating mantle 50ml round bottom flask,to 400 degrees C maximum internal operating temperature. 115V, 60W CSA. The Series TM mantle covers only the flask's bottom half, letting you see its full contents. Larger sizes have multiple circuits for ease of temperature control when a flask is less than half full. mantles are available with a bottom hole for flask drains.
Catalog No. 50-161-6364 Supplier Glas Col LLC Supplier No. 100B TM94
Aluminum housed heating mantle 50ml round bottom flask,to 400 degrees C maximum internal operating temperature. 115V, 60W CSA. The Series TM mantle covers only the flask's bottom half, letting you see its full contents. Larger sizes have multiple circuits for ease of temperature control when a flask is less than half full. mantles are available with a bottom hole for flask drains.