Learn More
Sigma Aldrich Fine Chemicals Biosciences Glas-ColR Series O hemisph1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z2847931EA
Encompass
Series O mantles are the original and the most popular heating mantles.Made from flexible fabricsfit easily into tight spacesMaximum 450 C internal operating temperatureAll mantles include a 4 ft 2-wire cord and locking connector. A temperature controller is required for operation. The mantles cover the bottom half of single and multi-neck flasks allowing for full view of contents.115 V models are CSA compliant230 V models are CE compliant
Catalog No. 50-290-0083