Learn More
Sigma Aldrich Fine Chemicals Biosciences Glas ColR Series O hemisph1EA
Catalog No. 502906035
Encompass
Series O mantles are the original and the most popular heating mantles.Made from flexible fabricsfit easily into tight spacesMaximum 450 C internal operating temperatureAll mantles include a 4 ft 2-wire cord and locking connector. A temperature controller is required for operation. The mantles cover the bottom half of single and multi-neck flasks allowing for full view of contents.115 V models are CSA compliant230 V models are CE compliant
Catalog No. 50-290-6035 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2848741EA
Series O mantles are the original and the most popular heating mantles.Made from flexible fabricsfit easily into tight spacesMaximum 450 C internal operating temperatureAll mantles include a 4 ft 2-wire cord and locking connector. A temperature controller is required for operation. The mantles cover the bottom half of single and multi-neck flasks allowing for full view of contents.115 V models are CSA compliant230 V models are CE compliant