Learn More
Sigma Aldrich Fine Chemicals Biosciences Glas ColR Series O spheric1EA
Catalog No. 502908850
Encompass
Help to improve safety and efficiency. Cover the top and bottom of 1 2 or 3-neck flask. Units 1L and larger have a secondary heating element in the top mantle. Not CE compliant.
Catalog No. 50-290-8850 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2850051EA
Help to improve safety and efficiency. Cover the top and bottom of 1 2 or 3-neck flask. Units 1L and larger have a secondary heating element in the top mantle. Not CE compliant.