Learn More
Sigma Aldrich Fine Chemicals Biosciences Glas ColR Series TM hemisp1EA
Catalog No. 502906059
Encompass
Series TM mantles have an aluminum exterior for extreme durability.Interior made from flexible fabricsMaximum 450C internal operating temperatureAll mantles include a 4ft 3-wire cord and locking connector. A temperature controller is required for operation. The mantle covers the bottom half of single and multi-neck flasks allowing for full view of contents.
Catalog No. 50-290-6059 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2852691EA
Series TM mantles have an aluminum exterior for extreme durability.Interior made from flexible fabricsMaximum 450C internal operating temperatureAll mantles include a 4ft 3-wire cord and locking connector. A temperature controller is required for operation. The mantle covers the bottom half of single and multi-neck flasks allowing for full view of contents.