Learn More
Sigma Aldrich Fine Chemicals Biosciences Glas ColR Series TM reacti1EA
Catalog No. 502905015
Encompass
Provides even heat for reaction kettles - ideal for resinous materials. Order heating mantle corresponding to flask manufacturer and size. Reaction flask not included.Not CE compliant
Catalog No. 50-290-5015 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2853661EA
Provides even heat for reaction kettles - ideal for resinous materials. Order heating mantle corresponding to flask manufacturer and size. Reaction flask not included.Not CE compliant