Learn More
Sigma Aldrich Fine Chemicals Biosciences Glas ColR Series TM reacti1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z2853821EA
Encompass
Provides even heat for reaction kettles - ideal for resinous materials. Order heating mantle corresponding to flask manufacturer and size. Reaction flask not included.Not CE compliant
Catalog No. 50-290-6036