Learn More
Glas Col LLC Heating Tape
SDP
Catalog No. NC0462145
Encompass_Preferred
This tape heats odd lengths of glass tubes and metal pipes. Ribbon-type heating elements running lengthwise in the tape are encased in durable glass braid and are impregnated and covered with tough silicone rubber, which insulates stranded terminal leads at one end and waterproofs fittings at both ends. Iideal for use in high moisture areas.. 480F Temperature Limit, 1/2" x 10'', 340W, 115Va
This item is not returnable. View return policy
Catalog No. NC0462145 Supplier Glas Col LLC Supplier No. 103A DET0.510
This tape heats odd lengths of glass tubes and metal pipes. Ribbon-type heating elements running lengthwise in the tape are encased in durable glass braid and are impregnated and covered with tough silicone rubber, which insulates stranded terminal leads at one end and waterproofs fittings at both ends. Iideal for use in high moisture areas.. 480F Temperature Limit, 1/2" x 10'', 340W, 115Va