Learn More
Andwin Scientific GLASCOL HEAT MANTEL NO0402
SDP
Catalog No. NC9336864
Encompass_Preferred
Glas-Col Flexible fabric heating mantles have insulated glass fabric that allow the vessel to conform into unusual shapes. Steel supports (sold separately) are needed to support mantle and allow attachment to ring stands. This product operates at 80 watts, with a flask size of 250 mL, max temperature of 842*F (450*C), and 115 VAC-DC.
Catalog No. NC9336864 Supplier Andwin Scientific Supplier No. 100A0402
Glas-Col Flexible fabric heating mantles have insulated glass fabric that allow the vessel to conform into unusual shapes. Steel supports (sold separately) are needed to support mantle and allow attachment to ring stands. This product operates at 80 watts, with a flask size of 250 mL, max temperature of 842*F (450*C), and 115 VAC-DC.