Learn More
Andwin Scientific Hemispherical Heating Mantle 1000mL, 18.1 O.D. x 9.7cm H, 450 deg.C
SDP
Catalog No. NC9096538
Encompass_Preferred
Hemispherical Heating Mantle, Glascol, bottom type O CSA 1000mL, 18.1 O.D. x 9.7cm H, 450 deg.C internal operating temp, Flask dia.13cm, 115V, 60Hz, 380w, Includes separable 1.2m, 2-wire cord, 1/EA
Catalog No. NC9096538 Supplier Andwin Scientific Supplier No. 100AO408
Hemispherical Heating Mantle, Glascol, bottom type O CSA 1000mL, 18.1 O.D. x 9.7cm H, 450 deg.C internal operating temp, Flask dia.13cm, 115V, 60Hz, 380w, Includes separable 1.2m, 2-wire cord, 1/EA