Learn More
TARGETMOL CHEMICALS INC GLASDEGIB 10MG
SDP
Catalog No. 502579341
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Glasdegib (PF-04449913) (PF-04449913) is a potent and orally bioavailable Smoothened (Smo) inhibitor with IC50 of 5 nM. Phase 2. purity: 98%
Catalog No. 50-257-9341 Supplier TARGETMOL CHEMICALS INC Supplier No. T651410MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Glasdegib (PF-04449913) (PF-04449913) is a potent and orally bioavailable Smoothened (Smo) inhibitor with IC50 of 5 nM. Phase 2. purity: 98%