Learn More
Medchemexpress LLC HY-16391 10mg Medchemexpress, Glasdegib CAS:1095173-27-5 Purity:>98%
SDP
Catalog No. 502030713
Encompass_Preferred
Medchemexpress, HY-16391 10mg Glasdegib CAS:1095173-27-5 Glasdegib (PF-04449913) is a potent and orally bioavailable smoothened inhibitor. Glasdegib (PF-04449913) binds to human SMO (amino acids 181-787) with an IC50 of 4 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0713 Supplier Medchemexpress LLC Supplier No. HY1639110MG
Medchemexpress, HY-16391 10mg Glasdegib CAS:1095173-27-5 Glasdegib (PF-04449913) is a potent and orally bioavailable smoothened inhibitor. Glasdegib (PF-04449913) binds to human SMO (amino acids 181-787) with an IC50 of 4 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.