Learn More
Sigma Aldrich Fine Chemicals Biosciences Glasgow Minimum Essential500ML
Catalog No. 502904613
Encompass
for use with adherent kidney cell lines such as baby hamster kidney cells (BHK).
Catalog No. 50-290-4613 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. G51546X500ML
for use with adherent kidney cell lines such as baby hamster kidney cells (BHK).