Learn More
Electron Microscopy Sciences Glasgow Washout Brush, Medium
SDP
Catalog No. 5030351
Encompass_Preferred
Four rows of white bristles locked in a wooden handle for the cleaning of the bench-top, around microtomes and microscopes etc. The overall length is 10".
Firmness: Medium
Catalog No. 50-303-51 Supplier Electron Microscopy Sciences Supplier No. 6558003
Four rows of white bristles locked in a wooden handle for the cleaning of the bench-top, around microtomes and microscopes etc. The overall length is 10".
Firmness: Medium