Learn More
Electron Microscopy Sciences Glasgow Washout Brush, Soft
SDP
Catalog No. 5030352
Encompass_Preferred
Four rows of white bristles locked in a wooden handle for the cleaning of the bench-top, around microtomes and microscopes etc. The overall length is 10".
Firmness: Soft
Catalog No. 50-303-52 Supplier Electron Microscopy Sciences Supplier No. 6558005
Four rows of white bristles locked in a wooden handle for the cleaning of the bench-top, around microtomes and microscopes etc. The overall length is 10".
Firmness: Soft