Learn More
West Pharmaceutical Services Inc Corning® Valor® Glass 10R RTU Vials with Stevanato Group EZ-fill® Technology, Nest & Tub. 720 vials/carton
Catalog No. NC3566831
Encompass
Valor® Glass vials with SG EZ-fill® technology are high-quality, ready-to-use vials for pharmaceutical and biotech drugs. The 10R ISO vials are compatible with West’s 20mm NovaPure® stoppers and 20mm Flip-Off® CCS seals (clean, certified, sterilized). These Valor® Glass vials are delivered in a nest & tub format. West item number: 100012585.
Catalog No. NC3566831 Supplier West Pharmaceutical Services Inc Supplier No. 100012585
Valor® Glass vials with SG EZ-fill® technology are high-quality, ready-to-use vials for pharmaceutical and biotech drugs. The 10R ISO vials are compatible with West’s 20mm NovaPure® stoppers and 20mm Flip-Off® CCS seals (clean, certified, sterilized). These Valor® Glass vials are delivered in a nest & tub format. West item number: 100012585.