Learn More
World Precision Instrument Multi-Barrel Capillary Glass-3B120F-4
SDP
Catalog No. 50821827
Encompass
Borosilicate glass capillaries: Close di­men­sion­al tol­er­anc­es assure mi­cro­elec­trode uni­for­mi­ty and reproducibility. Capillaries are available in 1, 2, 3, 5 and 7-barrel con­fig­u­ra­tions, complete range of single bar­rel thin-wall siz­es and a variety of special con­fig­u­ra­tions. Cap­il­lar­ies with filaments con­tain a solid fil­a­ment fused to the inner wall, which speeds filling of electrodes. Cap­il­lar­ies with or with­out inner filaments are available for making microelectrodes in a wide range of diameters.
Catalog No. 50-821-827 Supplier World Precision Instrument Supplier No. 3B120F4
Borosilicate glass capillaries: Close di­men­sion­al tol­er­anc­es assure mi­cro­elec­trode uni­for­mi­ty and reproducibility. Capillaries are available in 1, 2, 3, 5 and 7-barrel con­fig­u­ra­tions, complete range of single bar­rel thin-wall siz­es and a variety of special con­fig­u­ra­tions. Cap­il­lar­ies with filaments con­tain a solid fil­a­ment fused to the inner wall, which speeds filling of electrodes. Cap­il­lar­ies with or with­out inner filaments are available for making microelectrodes in a wide range of diameters.