Learn More
Chemglass Life Sciences Stirring Shaft, 10mm, Polished, Glass Agitator, Four-Blade, 460mm OAL, for 500mL Reactors, 67mm OD Agitator
SDP
Catalog No. CG2079B02
Encompass_Preferred
Stirring Shaft, 10mm, Clockwise, Polished, Glass Agitator, Three Blade, 400mm OAL, Agitator OD 42mm, for use with 100-250mL Reactors. Shaft is precision ground and polished to fit Chemglass's CG-2077-H, CG-2077-G and CG-2077-U style 10mm PTFE bearings. Designed to be used with cylindrical type reactors. The curved glass blades are spaced 120° apart and angled upward. PLEASE NOTE: STIRRER SHAFTS ARE DESIGNED FOR CLOCKWISE ROTATION.
Catalog No. CG2079B02 Supplier Chemglass Life Sciences Supplier No. CG2079B02
Stirring Shaft, 10mm, Clockwise, Polished, Glass Agitator, Three Blade, 400mm OAL, Agitator OD 42mm, for use with 100-250mL Reactors. Shaft is precision ground and polished to fit Chemglass's CG-2077-H, CG-2077-G and CG-2077-U style 10mm PTFE bearings. Designed to be used with cylindrical type reactors. The curved glass blades are spaced 120° apart and angled upward. PLEASE NOTE: STIRRER SHAFTS ARE DESIGNED FOR CLOCKWISE ROTATION.