Learn More
Chemglass Life Sciences Chemglass Glass Adapter, 1-1/2 in Beaded Pipe to 1 in Swagelok (for compression adapters)
SDP
Supplier:  Chemglass Life Sciences CG1964C100
Encompass_Preferred
Glass adapter designed to connect 1-1/2 in beaded glass pipe to a 1 in Swagelok compression fitting. It provides a sealed transition for fluid transfer between glass piping and Swagelok-style compression adapters, suitable for laboratory applications where glass-to-metal interfacing is required.
  • Connects 1-1/2 in beaded pipe to 1 in Swagelok compression fittings.
  • Fabricated from glass for chemical compatibility and visual monitoring.
  • Provides a secure, leak-resistant transition between glass and Swagelok connections.
  • Compatible with compression-style adapters and tubing systems.
Catalog No. 50-273-3560