Learn More
Chemglass Life Sciences Chemglass Glass Adapter, 1-1/2 in Beaded Pipe to 3/4 in Swagelok (for compression adapters)
SDP
Catalog No. 502733561
Encompass_Preferred
This glass adapter connects a 1-1/2 in beaded pipe to a 3/4 in Swagelok compression fitting for use in reactor and process glassware systems.
  • Connects 1-1/2 in beaded pipe to 3/4 in Swagelok compression fittings.
  • Made of glass for chemical compatibility.
  • Intended for use with compression adapters and reactor components.
  • Beaded pipe profile provides a seating surface for a secure seal when used with appropriate fittings.
Catalog No. 50-273-3561 Supplier Chemglass Life Sciences Supplier No. CG1964C075
This glass adapter connects a 1-1/2 in beaded pipe to a 3/4 in Swagelok compression fitting for use in reactor and process glassware systems.
  • Connects 1-1/2 in beaded pipe to 3/4 in Swagelok compression fittings.
  • Made of glass for chemical compatibility.
  • Intended for use with compression adapters and reactor components.
  • Beaded pipe profile provides a seating surface for a secure seal when used with appropriate fittings.