Learn More
Chemglass Life Sciences Glass Adapter, 1-1/2 in Beaded Pipe to CG-300-04 4-Ring Hose Connector
SDP
Catalog No. 502733559
Encompass_Preferred
This glass adapter provides a sealed transition between a 1-1/2 in beaded pipe and a 4-ring hose connector for reactor and process glassware systems. It is designed for fluid transfer and drainage connections where a secure hose termination is required.
  • Material: glass.
  • Connects a 1-1/2 in beaded pipe to a 4-ring hose connector.
  • Designed for use with process and reactor glassware hose connections.
  • Provides a secure, leak-resistant hose termination for fluid transfer and drainage.
  • Sold individually (each).
Catalog No. 50-273-3559 Supplier Chemglass Life Sciences Supplier No. CG1964G150
This glass adapter provides a sealed transition between a 1-1/2 in beaded pipe and a 4-ring hose connector for reactor and process glassware systems. It is designed for fluid transfer and drainage connections where a secure hose termination is required.
  • Material: glass.
  • Connects a 1-1/2 in beaded pipe to a 4-ring hose connector.
  • Designed for use with process and reactor glassware hose connections.
  • Provides a secure, leak-resistant hose termination for fluid transfer and drainage.
  • Sold individually (each).