Learn More
Ace Glass, Inc. Glass baffle, 12.7mm diax400mm length shaft. Fits #15Ace-Thred adapter. Baffle dimensions: 36mmx65mmx9.5mm, 465mm overall length.
SDP
Supplier:  Ace Glass, Inc. 1217719
Encompass
36mm X 65mm X 9.5mm Glass Baffle. Overall length: 465mm. Use with #15 Ace-Thred adapters to mate glass or PTFE (stainless steel core) baffles to any standard taper joint. Baffles can be adjusted vertically, or rotated to increase agitation.
Catalog No. 50-983-352