Learn More
Ace Glass, Inc. Glass baffle, 12.7mm diax50mm length shaft. Fits #15Ace-Thred adapter. Baffle dimensions-36mmx100mmx19.5mm, 600mm overall length.
SDP
Catalog No. 50977541
Encompass
36mm X 100mm X 19.5mm Glass Baffle. Overall length: 600mm. Use with #15 Ace-Thred adapters to mate glass or PTFE (stainless steel core) baffles to any standard taper joint. Baffles can be adjusted vertically, or rotated to increase agitation.
Catalog No. 50-977-541 Supplier Ace Glass, Inc. Supplier No. 1217723
36mm X 100mm X 19.5mm Glass Baffle. Overall length: 600mm. Use with #15 Ace-Thred adapters to mate glass or PTFE (stainless steel core) baffles to any standard taper joint. Baffles can be adjusted vertically, or rotated to increase agitation.