Learn More
Chemglass Life Sciences Glass Baffle Only for use in 10L-20L: Overall Length: 32", Baffle Width x Depth (mm): 20 x 30 Rectangle, Lower Style: Open
SDP
Catalog No. CG1872B20
Encompass_Preferred
Replacement Glass Baffle Only for use in 10L-20L. Overall Length: 32", Baffle Width x Depth (mm): 20 x 30 Rectangle, Lower Style: Open
Catalog No. CG1872B20 Supplier Chemglass Life Sciences Supplier No. CG1872B20
Replacement Glass Baffle Only for use in 10L-20L. Overall Length: 32", Baffle Width x Depth (mm): 20 x 30 Rectangle, Lower Style: Open