Learn More
Perkin Elmer US LLC Glass Baffled Cyclonic Spray Chamber with Gas Port for NexION 300/350 PC3 Systems
Catalog No. 502104879
Encompass_Preferred
This baffled cyclonic spray chamber is designed for all-purpose, non-hydrofluoric acid samples. The spray chamber has a secure, leak-free threaded drain port, O-ring-free connections to the nebulizer and injector, internal baffle and threaded auxiliary port for addition gas.
Catalog No. 50-210-4879 Supplier Perkin Elmer US LLC Supplier No. N8145119
This baffled cyclonic spray chamber is designed for all-purpose, non-hydrofluoric acid samples. The spray chamber has a secure, leak-free threaded drain port, O-ring-free connections to the nebulizer and injector, internal baffle and threaded auxiliary port for addition gas.