Learn More
Bellco Glass, Inc. Glass Ball Impeller Only,100mL
SDP
Catalog No. 501945121
Encompass_Preferred
Description: Glass Ball Impeller Assembly Only. MADE IN THE USASize: 100mLScrew Cap: Blue PP, 70mm OD x 22.4mm TCap Seal: Teflon, 67.9mm OD x 9.1mm TSealing Ring: Blue silicone rubber, 66.7mm OD x 47.6mm ID x 4.76mm TFlex Tube: Soft silicone, 41.3mm OAL, 0.312” OD x 0.248” IDMagnet Tube: Type 1 Class A Borosilicate Glass, 82mm OAL, 18mm OD ball, houses one cast alnico magnet
Catalog No. 50-194-5121 Supplier Bellco Glass, Inc. Supplier No. 196595010
Description: Glass Ball Impeller Assembly Only. MADE IN THE USASize: 100mLScrew Cap: Blue PP, 70mm OD x 22.4mm TCap Seal: Teflon, 67.9mm OD x 9.1mm TSealing Ring: Blue silicone rubber, 66.7mm OD x 47.6mm ID x 4.76mm TFlex Tube: Soft silicone, 41.3mm OAL, 0.312” OD x 0.248” IDMagnet Tube: Type 1 Class A Borosilicate Glass, 82mm OAL, 18mm OD ball, houses one cast alnico magnet