Learn More
Bellco Glass, Inc. Glass Ball Impeller Only,1L
SDP
Catalog No. 501945124
Encompass_Preferred
Description: Glass Ball Impeller Assembly Only. MADE IN THE USASize: 1000mLScrew Cap: Blue PP, 100mm OD x 22.4mm TCap Seal: Teflon, 95.3mm OD x 10.4mm TSealing Ring: Blue silicone rubber, 91.4mm OD x 76.2mm ID x 6.1mm TFlex Tube: Soft silicone, 76.2mm OAL, 0.312” OD x 0.248” IDMagnet Tube: Type 1 Class A Borosilicate Glass, 165mm OAL, 28mm OD ball, houses one cast alnico magnet
Catalog No. 50-194-5124 Supplier Bellco Glass, Inc. Supplier No. 196595100
Description: Glass Ball Impeller Assembly Only. MADE IN THE USASize: 1000mLScrew Cap: Blue PP, 100mm OD x 22.4mm TCap Seal: Teflon, 95.3mm OD x 10.4mm TSealing Ring: Blue silicone rubber, 91.4mm OD x 76.2mm ID x 6.1mm TFlex Tube: Soft silicone, 76.2mm OAL, 0.312” OD x 0.248” IDMagnet Tube: Type 1 Class A Borosilicate Glass, 165mm OAL, 28mm OD ball, houses one cast alnico magnet