Learn More
Bellco Glass, Inc. Glass Ball Spinner Complete,100mL
SDP
Catalog No. 501945107
Encompass_Preferred
•Material: Type 1 Class A Borosilicate Glass. MADE IN THE USA•Dimensions: 65mm Width x 135mm Overall Height•Graduations: 20mL – 100mL, every 20mL•Working Volume: 80-100mL•Specially Formed Bottom Indent Prevents Accumulation Under Impeller•Glass Ball Houses a Magnet for Gentle Agitation of Microcarrier Beads•5/16” OD x 1/4” ID Flex Tube Shaft•Two 32mm Side arms •One 70mm Screw Thread Center Neck
Catalog No. 50-194-5107 Supplier Bellco Glass, Inc. Supplier No. 196592001
•Material: Type 1 Class A Borosilicate Glass. MADE IN THE USA•Dimensions: 65mm Width x 135mm Overall Height•Graduations: 20mL – 100mL, every 20mL•Working Volume: 80-100mL•Specially Formed Bottom Indent Prevents Accumulation Under Impeller•Glass Ball Houses a Magnet for Gentle Agitation of Microcarrier Beads•5/16” OD x 1/4” ID Flex Tube Shaft•Two 32mm Side arms •One 70mm Screw Thread Center Neck